Skip to Content
Merck
Specific Gravity Hydrometer specific gravity 2.000-3.000 ( 0.010 subdivisions), L 305 mm

Specific Gravity Hydrometer

Specific Gravity Hydrometer specific gravity 2.000-3.000 ( 0.010 subdivisions), L 305 mm
Synonym(s):
densimeter, gravity meter, salinometer

Products

Product Category
Manufacturer
Showing 1-20 of 42
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3