Skip to Content
Merck
TAPI-2 TAPI-2, CAS 187034-31-7, is a hydroxamate-based inhibitor of MMPs and TACE. Inhibits the activation-induced shedding of L-selectin from neutrophils, eosinophils, and lymphocytes.

TAPI-2

TAPI-2 TAPI-2, CAS 187034-31-7, is a hydroxamate-based inhibitor of MMPs and TACE. Inhibits the activation-induced shedding of L-selectin from neutrophils, eosinophils, and lymphocytes.
Synonym(s):
TAPI-2, N-(R)-[2-(Hydroxyaminocarbonyl)methyl]-4-methylpentanoyl-L- t-butyl-alanyl-L-alanine, 2-aminoethyl Amide, TNF-α Protease Inhibitor-2, N-(R)-[2-(Hydroxyaminocarbonyl)methyl]-4-methylpentanoyl-L-t-butyl-alanyl-L-alanine, 2-aminoethyl Amide, TNF-α Protease Inhibitor-2
Empirical Formula (Hill Notation):
C19H37N5O5
CAS Number:
Molecular Weight:
415.53

Products

Product Category
Brand
Color
Physical Form
Purity
Research Area
Quality Segment