
TTNPB

Synonym(s):
4-[(E)-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid, Arotinoid acid
Empirical Formula (Hill Notation):
C24H28O2
CAS Number:
Molecular Weight:
348.48
Products
Product Category
Brand
Color
Physical Form
Purity
Quality Segment
Showing 1-2 of 2