Merck
  • Copper(II)- cis, cis-1,3,5-triaminocyclohexane complex-promoted hydrolysis of dipeptides: kinetic, speciation and structural studies.
Copper(II)- cis, cis-1,3,5-triaminocyclohexane complex-promoted hydrolysis of dipeptides: kinetic, speciation and structural studies.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2002-08-31)
Yuki Fujii, Tamas Kiss, Tamas Gajda, Xiang Shi Tan, Tsuyoshi Sato, Yoshiharu Nakano, Yoshihito Hayashi, Morio Yashiro
ABSTRACT
The hydrolysis of glycylglycine (GylGly), glycyl-L-leucine (GlyLeu), L-leucylglycine (LeuGly) and glycyl-DL-serine (GlySer) promoted by a copper(II)- cis, cis-1,3,5-triaminocyclohexane complex [Cu(II)TACH] was investigated at 70 degrees C and pH 7-10, using HPLC. The observed pseudo-first-order rate constants (k(obs)) and rate enhancing factors (REF) were as follows: 4.1x10(-3 )h(-1)(REF=23) for GylGly, 1.6x10(-3 )h(-1)(REF=21) for GlyLeu, 5.1x10(-3 )h(-1)(REF=64) for LeuGly and 9.2x10(-2 )h(-1)(REF=47) for GlySer [pH 8.1, dipeptide 2 mM, copper(II) 2 mM and TACH 2 mM]. Based on the pH dependence and dipeptide concentration dependence of the initial rates and speciation of the Cu(II)-TACH-dipeptide system at 25 degrees C and I=0.1, the reactions proceed via the formation of a ternary complex [Cu(TACH)(dipeptide)](+) as an intermediate followed by OH(-)-dependent and OH(-)-independent paths to give amino acid(s). GylGly, GlyLeu and LeuGly preferred the OH(-)-dependent path, while GlySer preferred the OH(-)-independent path. The latter can be explained by the intramolecular attack of the amide carbonyl group coordinated with its oxygen atom by the OH group in the serine residue. The X-ray crystal structure of [Cu(TACH)(GlyGly)]BPh(4).MeOH confirmed that GlyGly coordinates to copper(II) ion with its terminal amino N and amide O atoms. The crystal structures of [Cu(TACH)(Gly)]BPh(4) and [Cu(2)(TACH)(2)(OH)(2)](ClO(4))(2).NaClO(4).H(2)O are also reported.
Sigma-Aldrich
Leu-Gly
Pricing and availability is not currently available.