Merck
  • Simultaneous determination of tramadol and O-desmethyltramadol in human plasma using HPLC-DAD.
Simultaneous determination of tramadol and O-desmethyltramadol in human plasma using HPLC-DAD.
Journal of chromatographic science (2013-12-04)
Maha A Hilal, Khaled M Mohamed
ABSTRACT
A sensitive and accurate method for the extraction and quantification of tramadol (T) and its active metabolite, O-desmethyltramadol (ODT) in human plasma with high-performance liquid chromatography-diode array detection was developed and validated. The analytes were extracted from plasma samples by tert-butylmethyl ether in the presence of ammonium hydroxide as alkaline medium and back extraction with 1.0 M hydrochloric acid. Propranolol was used as internal standard. The extraction efficiencies of T and ODT were 83.51 and 78.72%, respectively. The calibration curves were linear (r(2) > 0.99) in the concentration range of 250-2000 ng/mL for T and ODT. Limits of detection and quantification were 125 and 250 ng/mL for both analytes. Intra- and interassay precision for T and ODT were ranged from 1.89 to 10.91% and 2.16 to 5.85%, respectively. Intra- and interassay accuracy for T and ODT were ranged from -13.07 to 4.99% and -2.03 to -6.98%, respectively. The method was successfully applied to quantify T and ODT from authentic plasma samples received from Hospital Sohag University. The method was completely validated and can be of interest to clinical and forensic laboratories.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Triethylamine, ≥99%
Supelco
Diethyl ether, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%