Merck
BAF378511000
CUBE 2UBE® Tubes
polypropylene, pack of 1000 tubes
About This Item
UNSPSC-Code:
41121800
Materialien
colorless tube
polypropylene
polypropylene tube
round bottom
Sterilität
non-sterile
Leistungsmerkmale
graduations: no
Verpackung
pack of 1000 tubes
Hersteller/Markenname
Bel-Art F37851-1000
Kapazität
2.2 mL
Bindungsart
non-treated surface
Allgemeine Beschreibung
CUBE 2UBE Tubes offer rapid, convenient sample storage and media preparation with 2.2ml per tube in a 8 x 12, 96 well format. The patented CUBE 2UBE Tubes is compatible with most automated liquid handling systems and can be centrifuged up to 14,000 RPM. The sturdy polypropylene CUBE 2UBE Tube can be frozen to -40 deg C. CUBE 2UBE Tubes are compatible with many Robotic workstations. Boxed in rigid polypropylene. Each tube has a capacity of 2.2MLS or 211MLS per rack of 96 tubes.
Rapid, convenient sample storage and media preparation in 2.2ml wells
  • Standard 8 x 12, 96 well format
  • Rugged polypropylene construction
Rechtliche Hinweise
CUBE 2UBE is a registered trademark of SP Scienceware, Division of SP