SAB1403544

Sigma-Aldrich

Monoclonal Anti-ALDOA antibody produced in mouse

clone 3C12-6D11, purified immunoglobulin, buffered aqueous solution

Synonym(e):
MGC17767, MGC17716, MGC10942, ALDA
NACRES:
NA.41
Preise und Verfügbarkeit sind derzeit nicht verfügbar.

Qualitätsniveau

100

Biologische Quelle

mouse

Antikörperform

purified immunoglobulin

antibody product type

primary antibodies

Klon

3C12-6D11, monoclonal

Form

buffered aqueous solution

Mol-Gew.

antigen ~66.15 kDa

species reactivity

human

Anwendung(en)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

Isotyp

IgG1κ

Konjugat

unconjugated

NCBI-Hinterlegungsnummer

UniProt-Hinterlegungsnummer

versandt in

dry ice

Lagertemp.

−20°C

Gene Information

human ... ALDOA(226)

Allgemeine Beschreibung

This gene product, Aldolase A (fructose-bisphosphate aldolase) is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Aldolase A is found in the developing embryo and is produced in even greater amounts in adult muscle. Aldolase A expression is repressed in adult liver, kidney and intestine and similar to aldolase C levels in brain and other nervous tissue. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. Alternative splicing of this gene results in multiple transcript variants which encode the same protein. (provided by RefSeq)

Immunogen

ALDOA (AAH10660.1, 1 a.a. ~ 364 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPYQYPALTPEQKKELSDIAHRIVAPGKGILAADESTGSIAKRLQSIGTENTEENRRFYRQLLLTADDRVNPCIGGVILFHETLYQKADDGRPFPQVIKSKGGVVGIKVDKGVVPLAGTNGETTTQGLDGLSERCAQYKKDGADFAKWRCVLKIGEHTPSALAIMENANVLARYASICQQNGIVPIVEPEILPDGDHDLKRCQYVTEKVLAAVYKALSDHHIYLEGTLLKPNMVTPGHACTQKFSHEEIAMATVTALRRTVPPAVTGITFLSGGQSEEEASINLNAINKCPLLKPWALTFSYGRALQASALKAWGGKKENLKAAQEEYVKRALANSLACQGKYTPSGQAGAAASESLFVSNHAY

Physikalische Form

Solution in phosphate buffered saline, pH 7.4

Haftungsausschluss

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Deutschland

WGK 3

Flammpunkt F

Not applicable

Flammpunkt C

Not applicable

Unser Team von Wissenschaftlern verfügt über Erfahrung in allen Forschungsbereichen einschließlich Life Science, Materialwissenschaften, chemischer Synthese, Chromatographie, Analytik und vielen mehr..

Setzen Sie sich mit dem technischen Dienst in Verbindung.

Soziale Medien

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Forschung. Entwicklung. Produktion.

Wir sind ein führender Anbieter für die globale Life-Science-Industrie mit Lösungen und Dienstleistungen für die Forschung, Entwicklung und Produktion in der Biotechnologie sowie für die Arzneimittelforschung und -produktion in der Pharmaindustrie.

© 2021 Merck KGaA, Darmstadt, Deutschland und/oder Tochterunternehmen. Alle Rechte vorbehalten.

Die Vervielfältigung von Inhalten dieser Internetseite ist ohne Genehmigung strengstens untersagt.