Iupac Name
2-hydrazino-1,1'-biphenyl hydrochloride
Inchi Code
1S/C12H12N2.ClH/c13-14-12-9-5-4-8-11(12)10-6-2-1-3-7-10;/h1-9,14H,13H2;1H
InChI key
XXKKAYKULLKATO-UHFFFAOYSA-N
storage temp.
Keep in the Freezer
Purity
96%
Country of Origin
US
Physical Form
Off White Solid
Product Link
https://www.jwpharmlab.com/home/product/20R0341S
MSDS
https://www.jwpharmlab.com/home/product/20R0341S
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service