Merck
All Photos(1)

4550-OP

Millipore

OmniPur® Ficoll® 400

form

powder

Quality Level

manufacturer/tradename

Calbiochem®

color

white

shipped in

ambient

General description

Ficoll® 400 is a neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution. Molecular biology grade.

Specifications:
DNase: None detected
Loss on drying: ≤5%
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Ficoll is a registered trademark of Cytiva
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service