Merck
All Photos(1)

06838

Sigma-Aldrich

Polysucrose 400-tetramethylrhodamine isothiocyanate conjugate

average mol wt 350,000-450,000

Synonym(s):
Tetramethylrhodamine isothiocyanate-polysucrose 400 conjugate, TRITC-Polysucrose 400
NACRES:
NA.32

form

powder

mol wt

average mol wt 350,000-450,000

fluorescence

λex 548 nm; λem 576 nm in 0.1 M phosphate pH 7.0

Analysis Note

degree of substitution 0.003-0.015

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Articles

FITC-labelled polysaccharides

Additional information about FITC-labelled polysaccarides including FITC-Dextran, FITC-DEAE-dextran (FDD), FITC-carboxymethyl-dextran (FCM-Dextran). This page discusses the impact of weight-average molecular weight on the respective molecule dimensions and other physical properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service