Merck
All Photos(1)

24914

Supelco

Purge/Trap E

for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Synonym(s):
PURGE TRAP E, Tekmar® 3000

application(s)

environmental

Quality Level

compatibility

for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials, so that a broad range of polar and nonpolar, high and low molecular weight compounds can be trapped in a single tube. Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition. During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds. During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents.

Packaging

1 ea

Legal Information

Tekmar is a registered trademark of Tekmar Co.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service