
Buffered Sodium Chloride Peptone Solution with Neutralizers

Products
Product Category
Brand
Physical Form
Agency
Chemical Composition
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing