<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
DE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Immobilon
®
-P PVDF Membrane
Products
Product Category
membranes (10)
sheets (8)
Brand
Millipore (10)
Length (cm)
Detection Method
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
IPVH00010
Immobilon
®
-P PVDF Membrane,
1 roll, 27 cm x 3.75 m, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH00005
Immobilon
®
-P PVDF Membrane,
1 roll, 27 cm x 1.875 m, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH10100
Immobilon
®
-P PVDF Membrane,
10 sheets, 10 cm x 10 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH15150
Immobilon
®
-P PVDF Membrane,
10 sheets, 15 cm x 15 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH09120
Immobilon
®
-P PVDF Membrane,
10 sheets, 9 cm x 12 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH07850
Immobilon
®
-P PVDF Membrane,
50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH20200
Immobilon
®
-P PVDF Membrane,
10 sheets, 20 cm x 20 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH304F0
Immobilon
®
-P PVDF Membrane,
10 sheets, 26 cm x 26 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH08100
Immobilon
®
-P PVDF Membrane,
10 sheets, 8 cm x 10 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
IPVH08130
Immobilon
®
-P PVDF Membrane,
10 sheets, 8.5 cm x 13.5 cm, 0.45 µm pore size, transfer membrane
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Deutsch - DE
English - EN
Learn More