Merck
Maltose monohydrate United States Pharmacopeia (USP) Reference Standard

Maltose monohydrate

Maltose monohydrate United States Pharmacopeia (USP) Reference Standard
Synonyms:
D-Maltose monohydrate, 4-O-α-D-Glucopyranosyl-D-glucose, D-(+)-Maltose monohydrate, Maltobiose, D-(+)-Maltose monohydrate, 4-O-α-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default