Skip to Content
DE
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silica gel 60 PF
254
containing gypsum
Synonym(s):
Silica gel
CAS Number:
7631-86-9
Products
Product Category
adsorbents (1)
Brand
Supelco (1)
Boiling Point (°C)
Melting Point (°C)
Physical Form
Quality Segment
Fluorescence
Matrix Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.07749
Silica gel 60 PF
254
containing gypsum,
for preparative layer chromatography
Expand
View Pricing