Merck
Tetrabutylammonium bisulfate solution ~55% in H2O

Tetrabutylammonium bisulfate solution

Tetrabutylammonium bisulfate solution ~55% in H2O
Synonyms:
Tetrabutylammonium hydrogen sulfate solution
Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53

Products

Shipping
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Package Size
Special Grade
Technique