Merck
  • Live attenuated Salmonella vaccines against Mycobacterium tuberculosis with antigen delivery via the type III secretion system.
Live attenuated Salmonella vaccines against Mycobacterium tuberculosis with antigen delivery via the type III secretion system.
Infection and immunity (2011-12-07)
María Dolores Juárez-Rodríguez, Lourdes T Arteaga-Cortés, Rebin Kader, Roy Curtiss, Josephine E Clark-Curtiss
ABSTRACT
Tuberculosis remains a global health threat, and there is dire need to develop a vaccine that is safe and efficacious and confers long-lasting protection. In this study, we constructed recombinant attenuated Salmonella vaccine (RASV) strains with plasmids expressing fusion proteins consisting of the 80 amino-terminal amino acids of the type 3 secretion system effector SopE of Salmonella and the Mycobacterium tuberculosis antigens early secreted antigenic target 6-kDa (ESAT-6) protein and culture filtrate protein 10 (CFP-10). We demonstrated that the SopE-mycobacterial antigen fusion proteins were translocated into the cytoplasm of INT-407 cells in cell culture assays. Oral immunization of mice with RASV strains synthesizing SopE-ESAT-6-CFP-10 fusion proteins resulted in significant protection of the mice against aerosol challenge with M. tuberculosis H37Rv that was similar to the protection afforded by immunization with Mycobacterium bovis bacillus Calmette-Guérin (BCG) administered subcutaneously. In addition, oral immunization with the RASV strains specifying these mycobacterial antigens elicited production of significant antibody titers to ESAT-6 and production of ESAT-6- or CFP-10-specific gamma interferon (IFN-γ)-secreting and tumor necrosis factor alpha (TNF-α)-secreting splenocytes.
Sigma-Aldrich
Monoclonal Anti-β-Galactosidase antibody produced in mouse, clone GAL-40, ascites fluid
Pricing and availability is not currently available.