Merck
  • Input pathways of organochlorine pesticides to typical freshwater cultured fish ponds of South China: hints for pollution control.
Input pathways of organochlorine pesticides to typical freshwater cultured fish ponds of South China: hints for pollution control.
Environmental toxicology and chemistry (2011-02-22)
Bao-Zhong Zhang, Huan-Yun Yu, Jing You, Eddy Y Zeng
ABSTRACT
Air, rain, pond water, bank soil, pond sediment, fish feed, and fish were sampled from four freshwater cultured fish ponds (FWCFPs) in rural areas within the Pearl River Delta (PRD) of South China. Compositional analyses indicated that historical residues were the main sources of DDXs (defined as the sum of dichlorodiphenyltrichloroethane (DDT), dichlorodiphenyldichloroethane (DDD), and dichlorodiphenyldichloroethylene (DDE) and 1-chloro-2,2-bis(4-chlorophenyl)ethane (p,p'-DDMU)), and hexachlorocyclohexanes (HCHs) in the FWCFPs. The input fluxes to the FWCFPs were estimated at 4.0, 1.6, 15, and -0.92 µg/m(2) ·year for DDXs and 3.8, 0.92, 2.9, and -1.4 µg/m(2) ·year for HCHs for dry deposition, wet deposition, feeding, and net air-water exchange in Dongguan, and 3.8, 1.2, 137, and -1.2 µg/m(2) ·year for DDXs and 3.6, 0.66, 5.0, and -1.0 µg/m(2) ·year for HCHs in Shunde, respectively. These results indicated that fish feed was the dominant input source of DDXs to the FWCFPs. As for HCHs, fluxes via dry deposition and feeding were similar and slightly higher than those via wet deposition. Biological effects due to the occurrence of DDXs in the FWCFPs were minimal, and consumption of freshwater fish from the PRD appeared to pose insignificant risk to human health based on some existing regulations and guidelines.