Merck
HPLC Analysis of Acidic and Neutral Drug Panel on Ascentis® Express C18
HPLC Analysis of Acidic and Neutral Drug Panel on Ascentis® Express C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
CONDITIONS
column
Ascentis Express C18, 10 cm × 4.6 mm I.D., 2.7 μm particles (53827-U)
mobile phase
[A] water with 0.1% phosphoric acid, [B] acetonitrile with 0.1% phosphoric acid
flow rate
1.76 mL/min
column temp.
ambient
detector
UV, 215 nm
injection
100 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany