Merck
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 2 mm
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 2 mm application for HPLC
CONDITIONS
column
Chromolith® Performance 100 x 2 mm (1.52006.0001)
mobile phase
[A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate
380 μL/min (u=2mm/s)
pressure
4619 psi (32 bar)
column temp.
ambient
detector
230 nm (cell: 1.4 μL)
injection
1.0 μL
sample
Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany