Merck
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 4 mm (analyte set 1
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 4 mm (analyte set 1 application for HPLC
CONDITIONS
column
Chromolith® Performance 100 x 4 mm (1.52028.0001)
mobile phase
[A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate
2 mL/min (u=2mm/s)
pressure
14001 psi (97 bar)
column temp.
ambient
detector
230 nm (cell: 1.4 μL)
injection
1 μL
sample
Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany