Merck
HPLC Analysis of Folic Acid Syrup on Chromolith® High Resolution RP-18
HPLC Analysis of Folic Acid Syrup on Chromolith® High Resolution RP-18 application for HPLC
CONDITIONS
sample preparation
Pipette out 15 ml of syrup in 50 ml volumetric flask. Add diluent to make up the volume.Sonicate for 30 min. Fiter through 0.2 μm filter paper.
column
Chromolith High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
mobile phase
[A] 1.5 g of hexane 1-sulfonate in 1000 ml water containing 10 ml of glacial acetic acid; [B] acetonitrile;
flow rate
1.0 mL/min
pressure
1203 psi 83 Bar
column temp.
30 °C
detector
277 nm cell: 10 μL
injection
10 μL
sample
10 ppm in diluent (Diluent: Dissolve 6.8 g of KH2PO4 in 1000 ml water. Adjust pH to 6.8 with 10% NaOH solution)
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany