Merck
HPLC Analysis of NSAIDs on Ascentis® RP-Amide
HPLC Analysis of NSAIDs on Ascentis® RP-Amide application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 10 mM dibasic potassium phosphate (pH 3.0): [B] acetonitrile; (45:55, A:B)
flow rate
1.0 mL/min.
column temp.
35 °C
detector
UV, 230 nm
injection
5 μL
sample
as indicated in 10 mM dibasic potassium phosphate (pH 3.0)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany