Merck
HPLC Analysis of NSAIDs on Discovery® Zr-Carbon C18
HPLC Analysis of NSAIDs on Discovery® Zr-Carbon C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Discovery® Zr-CarbonC18 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery Zr-Carbon C18, 15 cm × 4.6 mm I.D., 3 μm particles (65706-U)
mobile phase
[A] 50 mM phosphoric acid (pH 1.75): [B] acetonitrile; (50:50, A:B)
flow rate
4 mL/min
column temp.
80 °C
detector
UV, 254 nm
injection
1 μL
sample
ketoprofen, ibuprofen, naproxen, each 1 mg/mL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany