Merck
99100105
Opentrons® 200μL tip rack, filtered
new
Tips for AAW Workstation
Select a Size
About This Item
UNSPSC Code:
41121608
Volume:
200 μL
Sterility:
sterile
Feature:
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
Packaging:
pkg of 20 packs of 96 each
Manufacturer/tradename:
991-00105
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
feature
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
manufacturer/tradename
991-00105
packaging
pkg of 20 packs of 96 each
volume
200 μL
General description
200 µL filtered pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
Legal Information
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.
Visit the Document Library
Related Content
View an automated protocol for Duolink® assays on the AAW™ automated assay workstation and see results comparing manual vs automated runs.
Read an automated protocol for protein purification using PureProteome™ nickel magnetic beads on the AAW™ automated assay workstation and see results comparing manual vs automated runs.