Merck
All Photos(6)

ZRQSVR8WW

Milli-Q

Direct-Q® Water Purification System

flow rate 8 L/hr, A basic solution for pure and ultrapure water, equipped with a remote dispenser

Sign Into View Organizational & Contract Pricing
Synonym(s):
Direct-Q® 8 UV Remote Water Purification System

AC/DC input

100 - 230 V AC, 50 - 60 Hz (per your country requirements)

Quality Level

packaging

pack of 1 unit

parameter

12.2 kg operating weight (26.8 lb)
160 L/day max. usage (water)
8 L/hr flow rate

resistivity

18.2 MΩ-cm, at 25°C

H × W × D

54 cm (21.3 in.) × 29 cm (11.4 in.) × 38 cm (15 in.)

tank volume

6 L , Integrated storage tank

input

feed water nature potable tap water

output

product water quality: type 1 water (18.2 MΩ·cm)
product water quality: type 3 water (RO water)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRQSVR5WWSIMSV00WWZR0S0P8WW
parameter

12.2 kg operating weight (26.8 lb), 160 L/day max. usage (water), 8 L/hr flow rate

parameter

12.2 kg operating weight (26.8 lb), 5 L/hr flow rate, 60 L/day max. usage (water)

parameter

40 L/day max. usage (water), 9 kg operating weight (19.8 lb)

parameter

160 L/day max. usage (water), 8 L/hr flow rate, 8.9 kg operating weight (19.6 lb)

resistivity

18.2 MΩ-cm, at 25°C

resistivity

18.2 MΩ-cm, at 25°C

resistivity

18.2 MΩ-cm, at 25°C

resistivity

-

H × W × D

54 cm (21.3 in.) × 29 cm (11.4 in.) × 38 cm (15 in.)

H × W × D

54 cm (21.3 in.) × 29 cm (11.4 in.) × 38 cm (15 in.)

H × W × D

51 cm (20.1 in.) × 29 cm (11.4 in.) × 36 cm (14.2 in.)

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.0 cm (13.0 in.)

input

feed water nature potable tap water

input

feed water nature potable tap water

input

feed water nature pure water

input

feed water nature potable tap water

output

product water quality: type 1 water (18.2 MΩ·cm)
product water quality: type 3 water (RO water)

output

product water quality: type 1 water (18.2 MΩ·cm)
product water quality: type 3 water (RO water)

output

product water quality: type 1 water (18.2 MΩ·cm)

output

product water quality: type 3 water (RO water)

General description

The Direct-Q® 8 UV Remote Water Purification Systems deliver both pure and ultrapure water directly from the tap water for use in a wide variety of laboratory applications. Direct-Q® systems can be equipped with integrated (6 L) or external (30 or 60 L) water reservoirs and can be installed under or on the bench, or wall-mounted.
The system′s built-in 185 nm UV lamp reduces the levels of organics for critical applications. You can fine-tune your ultrapure water quality to match your applications using Application-specific point-of-use polishers to remove specific contaminants such as bacteria, particulates, pyrogen, nucleases, endocrine disruptors, or volatile organic compounds (VOCs).
The system′s versatile remote dispenser helps you stay focused on your work by providing ultrapure water delivery where you need it. The remote dispenser can be placed up to two meters from the Direct-Q® system and is available in free-standing or wall-mounted models.

Application

Pure water (Type 3) produced by the Direct-Q® 8 UV Remote Water Purification System is recommended for general laboratory tasks, preparation of buffers and reagent solutions, and rinsing of laboratory glassware.
Ultrapure water (Type 1) is used for analytical chemistry (liquid chromatography, elemental analysis, etc.), for molecular biology, biochemistry and cell culture media, for dissolving samples, etc.

Features and Benefits

Simple handling and maintenance:
  • Easy installation anywhere in your laboratory with its compact design and optional remote dispenser. Either wall-mounted, on or under the bench, choose the setup that is most convenient for your daily work.
  • Dispensing adapted to different size laboratory glassware at an instant flow rate of 0.5 L/min.
  • Save time thanks to the volumetric function. System automatically delivers the water volume that you request.
  • Easily insert your purification cartridge in one movement with the patented locking mechanism.

Components

  • This is a web reference. The end letters "WW" will be replaced by the country letters on your invoice to have the right shipping kit for the country. This ensures you receive the user manual in local language and a power cord adapted to the local electrical network.
  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

For more information on this product please contact our Technical Service Team

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Articles

Ultrapure Water for Perchlorate Analysis of Environmental Samples

Assure perchlorate-free water from tap water for sensitive IC analyses. See the removal efficiencies achieved by water purification technologies in Milli-Q® systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service