Merck
All Photos(1)

SAB2500233

Sigma-Aldrich

Anti-CHAT antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-B230380D24Rik, Anti-Choline acetyltransferase
MDL number:

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Chat(12647)

General description

Choline acetyltransferase (CHAT) has a molecular weight of about 68kDa. It is synthesized in the parakaryon and is transported to the axon terminals. CHAT is part of the choline/carnitine acyltransferase family.

Immunogen

Peptide with sequence C-KERARGPTKPSNLD from the C Terminus of the protein sequence according to NP_034021.1.

Application

Anti-CHAT antibody produced in goat has been used in immunohistochemistry.

Biochem/physiol Actions

Choline acetyltransferase (CHAT) is involved in the synthesis of acetylcholine in neurons. It catalyzes the transfer of an acetyl group between acetyl-coenzyme A and choline. Mutations in the gene encoding this protein have been associated with congenital neuromuscular disorders.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service