Merck
Sulfuric acid 95 - 97% for COD determination

Sulfuric acid

Sulfuric acid 95 - 97% for COD determination
Synonym(s):
Sulfuric acid, Sulfuric acid solution
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Physical Form
Product Line
Purity
Agency
Compatibility
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique