Skip to Content
Merck
Millistak+® HC Pod Depth Filter

Millistak+® HC Pod Depth Filter

Millistak+® HC Pod Depth Filter
Synonym(s):
Millistak+® HC Pro Pod Depth Filter

Products

Product Category
Brand
Sterilization Compatibility
Length (cm)
Quality Segment
Hydrophobicity
Application
Housing Material
Showing 1-20 of 41
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3