Skip to Content
Products
Cart
0
DK
EN
Products
Login / Register
Order Lookup
Quick Order
Cart
0
Back
T0901317
Synonym(s):
T0901317
Empirical Formula (Hill Notation):
C
17
H
12
NSO
3
F
9
CAS Number:
293754-55-9
Molecular Weight:
481.33
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
575310
T0901317,
≥98% (HPLC), liver X receptor agonist, solid
Expand
View Pricing
T2320
T0901317,
≥98%
Expand
View Pricing
An unknown error has occured.
English - EN
Learn More