Merck
  • Inhibition of lung tumor colonization and cell migration with the disintegrin crotatroxin 2 isolated from the venom of Crotalus atrox.
Inhibition of lung tumor colonization and cell migration with the disintegrin crotatroxin 2 isolated from the venom of Crotalus atrox.
Toxicon : official journal of the International Society on Toxinology (2008-04-05)
Jacob A Galán, Elda E Sánchez, Alexis Rodríguez-Acosta, Julio G Soto, Sajid Bashir, Mary Ann McLane, Carrie Paquette-Straub, John C Pérez
ABSTRACT
Disintegrins are low molecular weight proteins (4-15 kDa) with an RGD binding region at their binding loop. Disintegrin and disintegrin-like proteins are found in the venom of four families of snakes: Atractaspididae, Elapidae, Viperidae, and Colubridae. This report describes the biological activity of a disintegrin, crotatroxin 2, isolated by a three-step chromatography procedure from the venom of the Western diamondback rattlesnake (Crotalus atrox). The intact molecular mass for crotatroxin 2 was 7.384 kDa and 71 amino acids. Crotatroxin 2 inhibited human whole blood platelet aggregation with an IC(50) of 17.5 nM, inhibited cell (66.3p) migration by 63%, and inhibited experimental lung tumor colonization in BALB/c mice at 1000 microg/kg. Our data suggest that while crotatroxin 2 inhibits platelet aggregation, cancer cell migration, and lung tumor colonization, it is done via different integrins.
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg