Merck
  • Long-acting fentanyl analogues: synthesis and pharmacology of N-(1-phenylpyrazolyl)-N-(1-phenylalkyl-4-piperidyl)propanamides.
Long-acting fentanyl analogues: synthesis and pharmacology of N-(1-phenylpyrazolyl)-N-(1-phenylalkyl-4-piperidyl)propanamides.
Bioorganic & medicinal chemistry (2002-01-30)
Nadine Jagerovic, Carolina Cano, José Elguero, Pilar Goya, Luis F Callado, J Javier Meana, Rocío Girón, Raquel Abalo, David Ruiz, Carlos Goicoechea, M A Isabel Martín
ABSTRACT
The synthesis of new fentanyl analogues in which the benzene ring of the propioanilido group has been replaced by phenylpyrazole is described. Antinociceptive activity was evaluated using the writhing and hot plate tests in mice. Two compounds, and, showed interesting analgesic properties, being more potent than morphine and less than fentanyl but with longer duration of action. These compounds inhibited the electrically evoked muscle contraction of guinea pig ileum and mouse vas deferens but not that of rabbit vas deferens and the effects could be reversed by antagonists (naloxone and/or CTOP), thus indicating that the compounds acted as mu agonists. Finally, the binding data confirmed that the compounds had high affinity and selectivity for the mu receptor.