Merck
HomeWebinarsSPME Applications in Food Analysis by Means of a New Matrix Compatible Coating
SPME Applications in Food Analysis by Means of a New Matrix Compatible Coating
Webinar
Description / Abstract
The complexity and diversity food matrices and the need for clean extracts to be injected into analytical instrumentation may imply the use of tedious and time-consuming sample preparation strategies that often are a source of a significant amount of laboratory waste. The development of a new matrix-compatible solidphase microextraction (SPME) coating, namely PDMS/DVB/PDMS, bearing enhanced antifouling properties, enabled the analysis of complex food matrices by direct immersion SPME and helped to overcome issues related to extensive sample pre-treatment and instrumental contamination. This webinar will focus on recent advances made toward the analysis of contaminants in complex food samples by using the new matrix-compatible SPME coating and will describe the optimization of important parameters to be considered when performing DI-SPME.
Speaker
Emanuela Gionfriddo, Ph.D.
Research Associate under Prof. Janusz Pawliszyn, University of Waterloo (ON, Canada)
Webinar Information
Analytical chemistry
  • Sample preparation
Food and beverage testing and manufacturing
  • Chemical analysis
  • Microbiological analysis
  • Duration:65 mins
  • Language:English