Merck
Standard DNA Oligos
  • Scale: 0.025, 0.05, 0.2, 1.0, 10, and 15 μmol
  • Purification: Desalt, Cartridge, HPLC, and PAGE
  • Sequence Length: Up to 120 bases
  • Modifications: Over 200 available, including dyes (fluorescent and non-fluorescent)
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry or in solution – tubes, plates and mixed plates (forward and reverse primers)
  • WellRED Oligos: Available
  • qPCR Probes: Available
  • Learn more about Standard DNA Synthesis
Tubes
Plates
Easy Oligo
  • Amount: 3 OD
  • Concentration: 100 µM
  • Purification: Desalt
  • Sequence Length: 15 - 30 bases
  • Modifications: Not available
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied in solution
  • Ships within 24 hours of order receipt
  • Learn more
Tubes
Pure & Simple Primers
  • Amount: 3 OD
  • Purification: Cartridge
  • Sequence Length: 12 - 35 bases
  • Modifications: Not available
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry
  • Ships within 24 hours of order receipt
  • Learn more
Tubes
iScale Oligos™ (mg & g quantity)
  • Larger quantities for in vivo, high throughput and commercial projects
  • Quantity: 10, 25, 50, 100, 250, 500 mg; 1, 2, 5, 10, 15 g
  • Purification: Desalt, RP-HPLC & In vivo treatment
  • Sequence Length: 10 - 55 bases (Inquire for longer lengths)
  • Modifications: 6-FAM, Biotin, Phosphate, Amino-Modifiers & others
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry
  • Services: Salt Exchange, Filtration & Endotoxin Testing
  • Learn more
Tubes
Next-Gen Sequencing Oligos
  • Custom universal and index (barcode) adapter oligos for major sequencing platforms
  • Suitable purity (percent full-length sequence) to maintain sequence read integrity
  • Low levels of cross-contamination to facilitate proper sequence assembly
  • Learn more
Commercial Oligos
  • Custom DNA Primers, qPCR Probes, and Next-Gen Sequencing Oligos for Life Science Research Tools, Molecular Diagnostics (MDx), and Laboratory Developed Tests (LDTs)
  • Component or complete kit manufacturing, including custom formulations, packaging, and private labeling
  • Learn more
GMP OLIGOS
  • Custom cGMP Oligos, available with custom specifications, are Commercial Oligos (raw materials) for MDx and LDTs
  • Learn more
Locked Nucleic Acid
  • Available with Custom DNA Oligos
  • Increased thermal stability and hybridization specificity
  • Easier and more specific designs for problematic target sequences
  • Learn more
Tubes