Merck
All Photos(1)

GF37745275

Carbon - Vitreous

foil, 10mm disks, thickness 0.2mm, glassy carbon

Synonym(s):
Carbon - Vitreous, VC000250
Linear Formula:
C
CAS Number:
PubChem Substance ID:
NACRES:
NA.23

form

foil

manufacturer/tradename

Goodfellow 377-452-75

size diam. × thickness

10 mm × 0.2 mm

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service