Merck
All Photos(1)

F-025

Supelco

(±)-2-Fluoromethamphetamine-D5 (side chain) hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Empirical Formula (Hill Notation):
C10H9D5FN · HCl
Molecular Weight:
208.72
EC Number:

grade

certified reference material

Quality Level

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

FC1=C(C([2H])C(NC)([2H])C([2H])([2H])[2H])C=CC=C1.[H]Cl

InChI

1S/C10H14FN.ClH/c1-8(12-2)7-9-5-3-4-6-10(9)11;/h3-6,8,12H,7H2,1-2H3;1H/i1D3,7D,8D;

InChI key

GTSVNAIEHCXQPM-JRXSFSLASA-N

General description

A fluoro analog of methamphetamine, 2-Fluoromethamphetamine is sold in the illicit market as a club/designer drug. The illicit drug, also known as 2-FMA, produces stimulant and possibly empathogenic or psychoactive effects. This certified reference solution is suitable as an internal standard for GC/MS or LC/MS methamphetamine or amphetamine testing methods in clinical toxicology, forensic analysis or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service