Merck
All Photos(3)

ABN2294

Sigma-Aldrich

Anti-IQSEC3

new
Synonym(s):
IQ motif and SEC7 domain-containing protein 3;Potential synaptic guanine nucleotide exchange factor for Arf;SynArfGEF-Po;BRAG3

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 129.02 kDa
observed mol wt ~150 kDa

species reactivity

mouse, rat

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

Gene Information

Related Categories

General description

IQ motif and SEC7 domain-containing protein 3 (UniProt: Q76M68; also known as Potential synaptic guanine nucleotide exchange factor for Arf, SynArfGEF-Po) is encoded by the Iqsec3 (also known as Sag) gene (Gene ID: 404781) in rat. IQSEC3 along with IQSEC1 and IQSEC2 constitutes a family of brefeldin A-resistant ARF guanine nucleotide exchange factors (GEFs). It is a cytoplasmic protein that is expressed in the brain and is localized to dendrites and soma. Its expression is reported as early as E18 in hippocampal neurons. It is a key factor in controlling network activity in the hippocampus under both physiological and pathological conditions. It is shown to be exclusively localized to inhibitory synapses and it directly binds to gephyrin to promote inhibitory synapse formation in an ARF-GEF activity-dependent manner. IQSEC3 is reported to regulate GABAergic synapse maintenance in vivo and it links gephyrin-GABAA receptor complexes with ARF-dependent pathways to prevent aberrant, runaway excitation. Knockdown of Iqsec3 gene in the hippocampal dentate gyrus (DG) induces a significant reduction in GABAergic synapse density and causes a massive reduction of somatostatin expression in the DG hilus. IQ domain of IQSEC3 is localized to amino acids 311-340; the SEC7 domain is in amino acids 646-838; and the PH domain is in amino acids 852-985. (Ref.: Kim, S., et al. (2020). Cell Rep. 30(6); 1995-2005; Um, JW., et al. (2016). J. Biol. Chem. 291(19); 10119-10130).

Specificity

This rabbit polyclonal antibody detects IQ motif and SEC7 domain-containing protein 3 (IQSEC3). It targets an epitope within the N-terminal domain.

Immunogen

GST-tagged recombinant fragment corresponding to the first 185 amino acids from rat IQ motif and SEC7 domain-containing protein 3 (IQSEC3).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IQSEC3 in Rat brain tissue lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IQSEC3 in mouse brain tissue lysate.

Immunohistochemistry Applications: A representative lot detected IQSEC3 in Immunohistochemistry applications (Kim, S., et. al. (2020). Cell Rep. 30(6):1995-2005).

Immunocytochemistry Analysis: A representative lot detected IQSEC3 in Immunochemistry applications (Um, JW., et. al. (2016). J. Biol. Chem. 291(19):10119-30).

Immunoprecipitation Analysis: A representative lot detected IQSEC3 in Immunoprecipitation applications (Um, JW., et. al. (2016). J. Biol. Chem. 291(19):10119-30).

Western Blotting Analysis: A representative lot detected IQSEC3 in lysates from HEK293T cells transfected with Flag-IQSEC3, but not in untransfected cells. (Data Courtesy of Dr Jaewon Ko & Dr Seungjoon Kim at Daegu Gyeongbuk Institute of Science and Technology (DGIST)

Western Blotting Analysis: A representative lot detected IQSEC3 in Western Blotting applications (Kim, S., et. al. (2020). Cell Rep. 30(6):1995-2005).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IQSEC3, Cat. No. ABN2294, is a rabbit polyclonal antibody that detects IQSEC3 and is tested for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in 10 mM Tris-HCl pH 7.5 with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service