ZRB2107
Anti-RBPJ Antibody, clone 2D10 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Recombining binding protein suppressor of hairless;CBF-1;J kappa-recombination signal-binding protein;RBP-J kappa;RBP-J;RBP-JK;Renal carcinoma antigen NY-REN-30
Select a Size
Select a Size
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2D10, recombinant monoclonal
description
2D10 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 55.64 kDa
observed mol wt ~58 kDa
purified by
using Protein A
species reactivity
mouse, rat, human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... RBPJ(3516)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in NIH3T3 cell nuclear extract.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RBPJ in NIH3T3 cell nuclear extract.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RBPJ in HeLa cell nuclear extract.
Flow Cytometry Analysis: 0.1 μg from a representative lot detected RBPJ in one million Raji cells.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RBPJ in A431 and C6 cells.
Affinity Binding Assay: A representative lot of this antibody bound RBPJ peptide with a KD of 3.1 x 10-7 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected RBPJ in human stomach and human liver tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western Blotting in NIH3T3 cell nuclear extract.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RBPJ in NIH3T3 cell nuclear extract.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service