Products
ES
EN
Products
Applications
Products
Services
Support
Home
Search Results
16:0-18:1 PC
(2)
16:0-18:1 PC
Empirical Formula (Hill Notation):
C
42
H
82
NO
8
P
CAS Number:
26853-31-6
Molecular Weight:
760.08
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
850457P
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-
sn
-glycero-3-phosphocholine, powder
Expand
View Pricing
850457C
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-
sn
-glycero-3-phosphocholine, chloroform
Expand
View Pricing