Merck
L-Lysine monohydrochloride from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

(S)-(+)-Lysine monohydrochloride

L-Lysine monohydrochloride from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Synonyms:
(S)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, L-Lysine monohydrochloride, (S)-(+)-Lysine monohydrochloride
Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(S)-(+)-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
(S)-(+)-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
(S)-(+)-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
(S)-(+)-Lysine monohydrochloride, natural, FG
(S)-(+)-Lysine monohydrochloride, certified reference material, TraceCERT®
(S)-(+)-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
(S)-(+)-Lysine monohydrochloride, EMPROVE® ESSENTIAL Ph Eur,BP,USP
(S)-(+)-Lysine monohydrochloride, United States Pharmacopeia (USP) Reference Standard
(S)-(+)-Lysine monohydrochloride, European Pharmacopoeia (EP) Reference Standard
(S)-(+)-Lysine monohydrochloride, for synthesis