Merck
Ascentis® Express 160 Å C30, 2.7 μm HPLC Column column L × I.D. 5 cm × 3 mm

Ascentis® Express 160 Å C30, 2.7 μm HPLC Column

Ascentis® Express 160 Å C30, 2.7 μm HPLC Column column L × I.D. 5 cm × 3 mm
Synonym(s):
Ascentis® Express 160 Å C30, 2.7 μm HPLC Column

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique