Products
ES
EN
Products
Applications
Products
Services
Support
Home
Search Results
di-Sodium hydrogen phosphate anhydrous
(2)
di-Sodium hydrogen phosphate anhydrous
Synonyms:
Sodium hydrogenphosphate, Disodium phosphate, Disodium hydrogen phosphate, Sodium phosphate dibasic,
sec
-Sodium phosphate
Linear Formula:
Na
2
HPO
4
CAS Number:
7558-79-4
Molecular Weight:
141.96
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.06559
di-Sodium hydrogen phosphate anhydrous, for analysis particle size about 0.2-1 mm (~18-80 mesh ASTM) EMSURE
®
Expand
View Pricing
5.43838
di-Sodium hydrogen phosphate anhydrous, for HPLC LiChropur
™
Expand
View Pricing