Merck
  • Histomorphometric and immunohistochemical study of the goat rumen during prenatal development.
Histomorphometric and immunohistochemical study of the goat rumen during prenatal development.
Anatomical record (Hoboken, N.J. : 2007) (2012-02-23)
Angela García, Javier Masot, Antonio Franco, Antonio Gázquez, Eloy Redondo
ABSTRACT
This study sought to chart the ontogenesis of the goat rumen by histomorphometric examination, scanning electron microscopy and immunohistochemical analysis. A total of 140 goat embryos and fetuses were used, from the first stage of prenatal life until birth. The appearance of the rumen from the primitive gastric tube was observed at 35 days of prenatal life (CRL 3 cm, 23% gestation). By 38 days (CRL 4.3 cm CRL, 25% gestation) the ruminal wall comprised three layers: an internal epithelial layer, a middle layer of pluripotential blastemic tissue and an external layer or serosa. Ruminal pillars were visible at 46 days (CRL 6 cm, 30% gestation), and by 76 days (CRL 18 cm, 50% gestation) ruminal papillae were starting to appear. Under scanning electron microscopy, by 50 days (CRL 7.7 cm, 33% gestation) small ruminal papillae were observed protruding from the surface. Finally, neuroendocrine cells (synaptophysin, SYP) were detected at 53 days (CRL 9 cm CRL, 35% gestation), while glial cell markers (glial fibrillary acidic protein--GFAP, and vimentin-VIM) were found at 108 days (CRL 31 cm, 72% gestation) and 39 days (CRL 4.4 cm, 26% gestation), respectively. Neuropeptide Y (NPY) and vasoactive intestinal polypeptide (VIP) were detected immunohistochemically at 113 days (CRL 33 cm, 75% gestation) and 120 days (CRL 35 cm, 80% gestation), respectively. In conclusion, histomorphogenesis of the rumen in goats was similar to that reported in deer, but rather slower than observed for sheep or cattle.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution