Merck
  • Wnt6 expression in epidermis and epithelial tissues during Xenopus organogenesis.
Wnt6 expression in epidermis and epithelial tissues during Xenopus organogenesis.
Developmental dynamics : an official publication of the American Association of Anatomists (2008-01-29)
Danielle L Lavery, Ian R Davenport, Yvonne D Turnbull, Grant N Wheeler, Stefan Hoppler
ABSTRACT
Here, we report the localization within embryonic tissues of xWnt6 protein; together with the temporal and spatial expression of Xenopus laevis Wnt6 mRNA. Wnt6 expression in Xenopus embryos is low until later stages of neurulation, when it is predominantly found in the surface ectoderm. Wnt6 expression increases during early organogenesis in the epidermis overlaying several developing organs, including the eye, heart, and pronephros. At later stages of development, Wnt6 mRNA and protein generally localize in epithelial tissues and specifically within the epithelial tissues of these developing organs. Wnt6 localization correlates closely with sites of both epithelial to mesenchymal transformations and mesenchymal to epithelial transformations. Xenopus Wnt6 sequence and its expression pattern are highly conserved with other vertebrates. Xenopus embryos, therefore, provide an excellent model system for investigating the function of vertebrate Wnt6 in organ development and regulation of tissue architecture.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody