Merck
  • The Dolphin Proline-Rich Antimicrobial Peptide Tur1A Inhibits Protein Synthesis by Targeting the Bacterial Ribosome.
The Dolphin Proline-Rich Antimicrobial Peptide Tur1A Inhibits Protein Synthesis by Targeting the Bacterial Ribosome.
Cell chemical biology (2018-03-13)
Mario Mardirossian, Natacha Pérébaskine, Monica Benincasa, Stefano Gambato, Sven Hofmann, Paul Huter, Claudia Müller, Kai Hilpert, C Axel Innis, Alessandro Tossi, Daniel N Wilson
ABSTRACT
Proline-rich antimicrobial peptides (PrAMPs) internalize into susceptible bacteria using specific transporters and interfere with protein synthesis and folding. To date, mammalian PrAMPs have so far been identified only in artiodactyls. Since cetaceans are co-phyletic with artiodactyls, we mined the genome of the bottlenose dolphin Tursiops truncatus, leading to the identification of two PrAMPs, Tur1A and Tur1B. Tur1A, which is orthologous to the bovine PrAMP Bac7, is internalized into Escherichia coli, without damaging the membranes, using the inner membrane transporters SbmA and YjiL/MdM. Furthermore, like Bac7, Tur1A also inhibits bacterial protein synthesis by binding to the ribosome and blocking the transition from the initiation to the elongation phase. By contrast, Tur1B is a poor inhibitor of protein synthesis and may utilize another mechanism of action. An X-ray structure of Tur1A bound within the ribosomal exit tunnel provides a basis to develop these peptides as novel antimicrobial agents.
Sigma-Aldrich
Kanamycin sulfate, mixture of Kanamycin A (main component) and Kanamycin B and C
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%
Sigma-Aldrich
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)