Merck
  • Comparison of Protein Hydrolysis Catalyzed by Bovine, Porcine, and Human Trypsins.
Comparison of Protein Hydrolysis Catalyzed by Bovine, Porcine, and Human Trypsins.
Journal of agricultural and food chemistry (2018-04-05)
Yuxi Deng, Harry Gruppen, Peter A Wierenga
ABSTRACT
Based on trypsin specificity (for lysines and arginines), trypsins from different sources are expected to hydrolyze a given protein to the same theoretical maximum degree of hydrolysis (DHmax,theo). This is in contrast with experiments. Using α-lactalbumin and β-casein, this study aims to reveal if the differences among experimental DHmax (DHmax,exp) by bovine, porcine, and human trypsins are due to their secondary specificity. Peptide analysis showed that ∼78% of all the cleavage sites were efficiently hydrolyzed by porcine trypsin, and ∼47 and ∼53% were efficiently hydrolyzed by bovine and human trypsins, respectively. These differences were explained by the enzyme secondary specificity, that is, their sensitivities to the amino acids around the cleavage sites. The DHmax predictions based on the secondary specificity were 4 times closer to the DHmax,exp than the predictions based on trypsin specificity alone (DHmax,theo). Proposed preliminary relations between binding sites and trypsin secondary specificity allow DHmax,exp estimations of tryptic hydrolysis of other proteins.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
β-Casein from bovine milk, BioUltra, ≥98% (PAGE)
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein