- Bupivacaine-induced regeneration of rat soleus muscle: ultrastructural and immunohistochemical aspects.
Bupivacaine-induced regeneration of rat soleus muscle: ultrastructural and immunohistochemical aspects.
Ultrastructural Pathology (2006)
Politi PK, et al.