Merck
  • Fully automated standard addition method for the quantification of 29 polar pesticide metabolites in different water bodies using LC-MS/MS.
Fully automated standard addition method for the quantification of 29 polar pesticide metabolites in different water bodies using LC-MS/MS.
Analytical and Bioanalytical Chemistry (2013)
Kowal, et al.