Merck
  • Abolishment of the interaction between cyclin-dependent kinase 2 and Cdk-associated protein phosphatase by a truncated KAP mutant.
Abolishment of the interaction between cyclin-dependent kinase 2 and Cdk-associated protein phosphatase by a truncated KAP mutant.
Biochemical and biophysical research communications (2003-05-15)
Chau-Ting Yeh, Su-Chuan Lu, Chung-Hao Chao, Mei-Ling Chao
ABSTRACT
The cyclin-dependent kinase (Cdk)-associated protein phosphatase (KAP) is a human dual-specificity protein phosphatase that dephosphorylates Cdk2 on a conserved threonine residue, T160, in a cyclin dependent manner. Several aberrant KAP transcripts with characteristic deletion regions have been identified in hepatocellular carcinoma tissues. In this report, we demonstrated that multiple aberrant KAP transcripts were also present in a hepatoblastoma cell line (HepG2), albeit harboring a totally different set of deletions. By performing yeast two-hybrid and co-immunoprecipitation experiments, a KAP-Cdk2 interaction domain located in the amino acid 1-34 region was identified. This interaction domain was different from the major protein interface deduced from crystal structure analysis. Using a yeast three-hybrid system, it was shown that the presence of a truncated KAP mutant encoding this interaction domain abolished the wild-type KAP-Cdk2 interaction. In conclusion, a previously unidentified KAP-Cdk2 interaction domain was discovered. Truncated KAP mutants containing this domain interfered with the wild-type KAP-Cdk2 interaction.
Sigma-Aldrich
CDKN3, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution