Merck
  • Determination of inorganic cations and ammonium in environmental waters by ion chromatography with a high-capacity cation-exchange column
Determination of inorganic cations and ammonium in environmental waters by ion chromatography with a high-capacity cation-exchange column
Journal of Chromatography A (2002)
Thomas DH, et al.